Product Details

SNP ID
rs277640
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:100368017 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCTAAATATCTATGGAGCTACCATA[C/T]ATTTCCTAACAGACTCCAGAAGTTA
Phenotype
MIM: 606081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TOMM70 PubMed Links

Gene Details

Gene
TOMM70
Gene Name
translocase of outer mitochondrial membrane 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014820.4 Intron NP_055635.3

View Full Product Details