Product Details

SNP ID
rs1437283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:1312521 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCCAACACTTGCAGCCCCTCCA[A/G]AGAAAAAGCCCAGCGGAAGAGGACT
Phenotype
MIM: 612788
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXQ1 PubMed Links

Gene Details

Gene
FOXQ1
Gene Name
forkhead box Q1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033260.3 82 UTR 5 NP_150285.3
Gene
LINC01394
Gene Name
long intergenic non-protein coding RNA 1394
There are no transcripts associated with this gene.

Gene
LOC107986554
Gene Name
uncharacterized LOC107986554
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017011607.1 82 Intron XP_016867096.1
XM_017011608.1 82 Intron XP_016867097.1
XM_017011609.1 82 Intron XP_016867098.1
XM_017011610.1 82 Intron XP_016867099.1
XM_017011611.1 82 Intron XP_016867100.1

View Full Product Details