Product Details

SNP ID
rs1044239
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135933074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCTACCTCCGTCAAATAACAAG[G/T]GGACTCTCCAAGCAAATGTCTACAC
Phenotype
MIM: 608129
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
UBAC1 PubMed Links
Additional Information
For this assay, SNP(s) [rs139960127] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
UBAC1
Gene Name
UBA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016172.2 1762 UTR 3 NP_057256.2

View Full Product Details