Product Details

SNP ID
rs1061733
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:158733970 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGAGTCCACAGACACAAGAGCTAC[C/T]GCCGCCCTACTGCACGTTGTAGGAA
Phenotype
MIM: 616691
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESYT2 PubMed Links

Gene Details

Gene
ESYT2
Gene Name
extended synaptotagmin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020728.2 2985 UTR 3 NP_065779.1

View Full Product Details