Product Details

SNP ID
rs1048179
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:95245183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTAGAAACTATACTTTTCTTCTA[C/T]ACTGATACTAAAATAGTTTCATTTT
Phenotype
MIM: 614477
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8orf37 PubMed Links

Gene Details

Gene
C8orf37
Gene Name
chromosome 8 open reading frame 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177965.3 4375 UTR 3 NP_808880.1
XM_005250799.3 4375 UTR 3 XP_005250856.2

View Full Product Details