Product Details

SNP ID
rs1142283
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:23434322 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCCAAATTCACAGCCCTACCTTAA[C/T]GGTAGAAAGCGGGTCCTGTAGAGGA
Phenotype
MIM: 607577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENTPD4 PubMed Links

Gene Details

Gene
ENTPD4
Gene Name
ectonucleoside triphosphate diphosphohydrolase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128930.2 1865 Silent Mutation CCA,CCG P531P NP_001122402.1
NM_004901.4 1865 Silent Mutation CCA,CCG P539P NP_004892.1

View Full Product Details