Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001284338.1 | 2868 | UTR 3 | NP_001271267.1 | ||
NM_001284339.1 | 2868 | UTR 3 | NP_001271268.1 | ||
NM_001284340.1 | 2868 | UTR 3 | NP_001271269.1 | ||
NM_006154.3 | 2868 | UTR 3 | NP_006145.2 | ||
NM_198400.3 | 2868 | UTR 3 | NP_940682.2 | ||
XM_011521624.2 | 2868 | UTR 3 | XP_011519926.1 | ||
XM_011521625.2 | 2868 | UTR 3 | XP_011519927.1 | ||
XM_011521626.1 | 2868 | UTR 3 | XP_011519928.1 | ||
XM_011521627.1 | 2868 | UTR 3 | XP_011519929.1 |