Product Details

SNP ID
rs1045741
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:107733496 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACAAAGATCTCCACGTTACAATAA[C/G]CAGGTGACTTGCTCAGGCTTCCATC
Phenotype
MIM: 605780
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929162 PubMed Links

Gene Details

Gene
LOC101929162
Gene Name
uncharacterized LOC101929162
There are no transcripts associated with this gene.

Gene
PRDM4
Gene Name
PR domain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012406.3 3557 UTR 3 NP_036538.3
XM_005268593.1 3557 UTR 3 XP_005268650.1

View Full Product Details