Product Details

SNP ID
rs1702832
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:75196338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTGGAGGCACATGTTCAAATTT[C/T]TGCACTTAGGCCAGTATGGTGGCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZADH2 PubMed Links

Gene Details

Gene
ZADH2
Gene Name
zinc binding alcohol dehydrogenase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306093.1 6324 UTR 3 NP_001293022.1
NM_175907.5 6324 UTR 3 NP_787103.1

View Full Product Details