Product Details
- SNP ID
-
rs1519463
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
52
- Location
-
Chr.1:221704797 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATCACCCTGAATTTCCTTTCAGAC[C/T]CAATATTCCATGGTCCTGTGCTGAC
- Phenotype
-
MIM: 608867
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DUSP10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116977823] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DUSP10
- Gene Name
- dual specificity phosphatase 10
View Full Product Details