Product Details

SNP ID
rs949994
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:220881037 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTTTTCGTGCGTCGCTCCTTGGC[A/G]ACTGTTGGGAGAGAGAGGTTTCAGC
Phenotype
MIM: 142995
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLX PubMed Links

Gene Details

Gene
HLX
Gene Name
H2.0 like homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021958.3 Intron NP_068777.1
Gene
HLX-AS1
Gene Name
HLX antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details