Product Details

SNP ID
rs1028558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:141607108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCAATTAGAGGCCCTAATTGCAA[G/T]GTCATCCCCAGCTCCACCCTGCTCC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPANXA2-OT1 PubMed Links

Gene Details

Gene
SPANXA2-OT1
Gene Name
SPANXA2 overlapping transcript 1
There are no transcripts associated with this gene.

View Full Product Details