Product Details

SNP ID
rs916400
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44676634 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCGGGGTGAAGAGGAGGAGGGGGG[C/T]TGTCCAAGGGGCATAGCAGTCGGAG
Phenotype
MIM: 609406
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PRR5 PubMed Links
Additional Information
For this assay, SNP(s) [rs79134873] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
PRR5
Gene Name
proline rich 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017528.2 Intron NP_001017528.1
NM_001017529.2 Intron NP_001017529.1
NM_001017530.1 Intron NP_001017530.1
NM_001198721.1 Intron NP_001185650.1
NM_015366.3 Intron NP_056181.2
NM_181333.3 Intron NP_851850.1

View Full Product Details