Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_033159.3 | 2638 | UTR 3 | NP_149349.2 | ||
NM_153281.1 | 2638 | UTR 3 | NP_695013.1 | ||
NM_153282.2 | 2638 | UTR 3 | NP_695014.1 | ||
NM_153283.2 | 2638 | UTR 3 | NP_695015.1 | ||
NM_153285.2 | 2638 | UTR 3 | NP_695017.1 | ||
XM_011533667.2 | 2638 | UTR 3 | XP_011531969.1 | ||
XM_011533668.2 | 2638 | UTR 3 | XP_011531970.1 | ||
XM_011533669.2 | 2638 | UTR 3 | XP_011531971.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001200029.1 | 2638 | Intron | NP_001186958.1 | ||
NM_001200030.1 | 2638 | Intron | NP_001186959.1 | ||
NM_001200031.1 | 2638 | Intron | NP_001186960.1 | ||
NM_001200032.1 | 2638 | Intron | NP_001186961.1 | ||
NM_003549.3 | 2638 | Intron | NP_003540.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001200016.1 | 2638 | Intron | NP_001186945.1 | ||
NM_001200018.1 | 2638 | Intron | NP_001186947.1 | ||
NM_012191.3 | 2638 | Intron | NP_036323.2 |