Product Details

SNP ID
rs10523
Assay Type
Validated
NCBI dbSNP Submissions
69
Location
Chr.1:6635184 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
AGATTCAAGCAGTAATATTTCAGCA[A/G]GAGGAGGTGGTCATGGGAAGCCAGC
Phenotype
MIM: 614827 MIM: 612532
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC11 PubMed Links

Gene Details

Gene
DNAJC11
Gene Name
DnaJ heat shock protein family (Hsp40) member C11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018198.3 1630 UTR 3 NP_060668.2
XM_017001745.1 1630 UTR 3 XP_016857234.1
Gene
THAP3
Gene Name
THAP domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195752.1 1630 Intron NP_001182681.1
NM_001195753.1 1630 Intron NP_001182682.1
NM_138350.3 1630 UTR 3 NP_612359.2
XM_005263532.3 1630 Intron XP_005263589.1
XM_011542400.2 1630 Intron XP_011540702.1
XM_011542401.2 1630 Intron XP_011540703.1
XM_017002761.1 1630 UTR 3 XP_016858250.1

View Full Product Details