Product Details

SNP ID
rs704244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48813083 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGCACCTTGCTAACTGGGCTTA[A/G]AGTCTAAGGGCTTGGGGGCTGCATC
Phenotype
MIM: 113530 MIM: 612832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCAT2 PubMed Links

Gene Details

Gene
BCAT2
Gene Name
branched chain amino acid transaminase 2
There are no transcripts associated with this gene.

Gene
HSD17B14
Gene Name
hydroxysteroid 17-beta dehydrogenase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016246.2 1171 UTR 3 NP_057330.2
XM_005258969.3 1171 UTR 3 XP_005259026.1
XM_005258970.3 1171 UTR 3 XP_005259027.1

View Full Product Details