Product Details
- SNP ID
-
rs1449683
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:122826931 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGCGGCGTCCGCGGAGACACCCATC[C/T]GTGAACCCCAGGTCCCGGGCCGCCG
- Phenotype
-
MIM: 134920
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FGF2
PubMed Links
Gene Details
- Gene
- FGF2
- Gene Name
- fibroblast growth factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002006.4 |
224 |
Silent Mutation |
TCC,TCT |
S52S |
NP_001997.5 |
View Full Product Details