Product Details

SNP ID
rs817317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63954200 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACCACTCAGGAAACCTTCCCTC[A/T]GCAGGACCTGCCCTGCACAGATCAT
Phenotype
MIM: 610866
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UCKL1 PubMed Links

Gene Details

Gene
UCKL1
Gene Name
uridine-cytidine kinase 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193379.1 Intron NP_001180308.1
NM_017859.3 Intron NP_060329.2
XM_005260216.2 Intron XP_005260273.1
XM_006723806.2 Intron XP_006723869.1
XM_006723807.2 Intron XP_006723870.1
XM_006723808.1 Intron XP_006723871.1
XM_006723809.1 Intron XP_006723872.1
XM_006723810.1 Intron XP_006723873.1
XM_006723811.3 Intron XP_006723874.1
XM_011528868.2 Intron XP_011527170.1
XM_011528869.1 Intron XP_011527171.1
XM_011528870.1 Intron XP_011527172.1
XM_011528871.1 Intron XP_011527173.1
XM_011528872.1 Intron XP_011527174.1
XM_011528873.2 Intron XP_011527175.1
XM_017027894.1 Intron XP_016883383.1
XM_017027895.1 Intron XP_016883384.1
XM_017027896.1 Intron XP_016883385.1
XM_017027897.1 Intron XP_016883386.1
XM_017027898.1 Intron XP_016883387.1
XM_017027899.1 Intron XP_016883388.1
XM_017027900.1 Intron XP_016883389.1
XM_017027901.1 Intron XP_016883390.1
XM_017027902.1 Intron XP_016883391.1
XM_017027903.1 Intron XP_016883392.1
XM_017027904.1 Intron XP_016883393.1
XM_017027905.1 Intron XP_016883394.1
XM_017027906.1 Intron XP_016883395.1
XM_017027907.1 Intron XP_016883396.1
XM_017027908.1 Intron XP_016883397.1
Gene
UCKL1-AS1
Gene Name
UCKL1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
ZNF512B
Gene Name
zinc finger protein 512B
There are no transcripts associated with this gene.

View Full Product Details