Product Details

SNP ID
rs975915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:73976658 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAAGTAGCAGGCCGAGGGCCCC[C/G]GGGTCCTCAGCCTCCGCGAGTCCAG
Phenotype
MIM: 600788 MIM: 612418
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TCEB1 PubMed Links

Gene Details

Gene
TCEB1
Gene Name
transcription elongation factor B subunit 1
There are no transcripts associated with this gene.

Gene
TMEM70
Gene Name
transmembrane protein 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040613.2 Intron NP_001035703.1
NM_017866.5 Intron NP_060336.3

View Full Product Details