Product Details

SNP ID
rs1460249
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:75412744 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATTTTTTTTTTTGCATTTTCTTATT[G/T]CAGACATCATAGTTACAGGTTCCAA
Phenotype
MIM: 605966
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HNF4G PubMed Links

Gene Details

Gene
HNF4G
Gene Name
hepatocyte nuclear factor 4 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004133.4 Intron NP_004124.4
XM_011517516.2 Intron XP_011515818.1
XM_017013373.1 Intron XP_016868862.1
XM_017013374.1 Intron XP_016868863.1
XM_017013375.1 Intron XP_016868864.1
XM_017013376.1 Intron XP_016868865.1

View Full Product Details