Product Details

SNP ID
rs1415793
Assay Type
Functionally tested
NCBI dbSNP Submissions
78
Location
Chr.1:111499614 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCACACACATGTTCAGCCCAACT[A/G]GAGCCTTTTGTCAGTAAGTCAACTA
Phenotype
MIM: 600445
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADORA3 PubMed Links

Gene Details

Gene
ADORA3
Gene Name
adenosine A3 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000677.3 2060 UTR 3 NP_000668.1
NM_001302678.1 2060 UTR 3 NP_001289607.1
NM_001302679.1 2060 UTR 3 NP_001289608.1
Gene
TMIGD3
Gene Name
transmembrane and immunoglobulin domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081976.2 2060 Intron NP_001075445.1
NM_001302680.1 2060 Intron NP_001289609.1
NM_020683.6 2060 Intron NP_065734.5

View Full Product Details