Product Details

SNP ID
rs2007683
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:36133785 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATAATTGTAGAATGTGGTAAGATAG[C/T]ATTGATTCAGATATCAACATGAAAG
Phenotype
MIM: 602899
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAPK13 PubMed Links

Gene Details

Gene
MAPK13
Gene Name
mitogen-activated protein kinase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002754.4 Intron NP_002745.1

View Full Product Details