Product Details

SNP ID
rs1557153
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:218345918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCCCGCCAGGGTGTAGGCCACGG[A/C]GCGCAGCTCCCAGAGCAGGATCCGC
Phenotype
MIM: 611193 MIM: 190220
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RRP15 PubMed Links

Gene Details

Gene
RRP15
Gene Name
ribosomal RNA processing 15 homolog
There are no transcripts associated with this gene.

Gene
TGFB2
Gene Name
transforming growth factor beta 2
There are no transcripts associated with this gene.

Gene
TGFB2-AS1
Gene Name
TGFB2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details