Product Details

SNP ID
rs1376426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:65321743 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAACTTAAAGTTCTAGTCATTTA[A/G]ATACACTCCACCCGGACCTCCTAAT
Phenotype
MIM: 600004
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPHA5 PubMed Links

Gene Details

Gene
EPHA5
Gene Name
EPH receptor A5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281765.2 6026 UTR 3 NP_001268694.1
NM_001281766.2 6026 UTR 3 NP_001268695.1
NM_001281767.2 6026 Intron NP_001268696.1
NM_001318761.1 6026 UTR 3 NP_001305690.1
NM_004439.7 6026 UTR 3 NP_004430.4
NM_182472.4 6026 UTR 3 NP_872272.2
XM_005265653.3 6026 UTR 3 XP_005265710.1
XM_011531735.2 6026 UTR 3 XP_011530037.1
XM_017007878.1 6026 UTR 3 XP_016863367.1
XM_017007879.1 6026 Intron XP_016863368.1
XM_017007880.1 6026 UTR 3 XP_016863369.1
XM_017007881.1 6026 Intron XP_016863370.1

View Full Product Details