Product Details

SNP ID
rs9347
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:182384063 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CCCTTTCAGCTACCTGGAAGAAGAG[A/T]CCCCCTTCTGCAAACGTGCTCTGTC
Phenotype
MIM: 138290
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GLUL PubMed Links

Gene Details

Gene
GLUL
Gene Name
glutamate-ammonia ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033044.3 2068 UTR 3 NP_001028216.1
NM_001033056.3 2068 UTR 3 NP_001028228.1
NM_002065.6 2068 UTR 3 NP_002056.2
XM_006711278.1 2068 UTR 3 XP_006711341.1

View Full Product Details