Product Details

SNP ID
rs885828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:71818746 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCTATATGGTGAGAAAAGGAAA[A/G]AAGAAAGGGGGAGAATGAGAGCTGC
Phenotype
MIM: 605516 MIM: 176801
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDH23 PubMed Links

Gene Details

Gene
CDH23
Gene Name
cadherin-related 23
There are no transcripts associated with this gene.

Gene
PSAP
Gene Name
prosaposin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042465.2 Intron NP_001035930.1
NM_001042466.2 Intron NP_001035931.1
NM_002778.3 Intron NP_002769.1

View Full Product Details