Product Details

SNP ID
hCV8867589
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:82822013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGAAAAAAATTACATTAAAAAAA[G/C]AAGTTAATCTGGAAGGATGCATGCC
Phenotype
MIM: 612393
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
WHAMM PubMed Links

Gene Details

Gene
WHAMM
Gene Name
WAS protein homolog associated with actin, golgi membranes and microtubules
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080435.2 Intron NP_001073904.1

View Full Product Details