Product Details

SNP ID
rs1048205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:39799371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGATGCCATGCACGCAGGGCCCA[A/G]AAGGCAGCAGAGCCGCAGGAGGCTG
Phenotype
MIM: 612183
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR176 PubMed Links

Gene Details

Gene
GPR176
Gene Name
G protein-coupled receptor 176
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271854.1 4498 UTR 3 NP_001258783.1
NM_001271855.1 4498 UTR 3 NP_001258784.1
NM_007223.2 4498 UTR 3 NP_009154.1
XM_011521167.2 4498 UTR 3 XP_011519469.1
XM_011521168.2 4498 UTR 3 XP_011519470.1
XM_017021873.1 4498 UTR 3 XP_016877362.1
XM_017021874.1 4498 UTR 3 XP_016877363.1
XM_017021875.1 4498 UTR 3 XP_016877364.1
XM_017021876.1 4498 UTR 3 XP_016877365.1
XM_017021877.1 4498 UTR 3 XP_016877366.1
XM_017021878.1 4498 UTR 3 XP_016877367.1

View Full Product Details