Product Details

SNP ID
rs7875
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:137752360 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAACCATGTCAAGATGATAAACAA[C/T]TCGCTTAGTACAAGAGATGGATGCA
Phenotype
MIM: 609409
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPA0 PubMed Links

Gene Details

Gene
HNRNPA0
Gene Name
heterogeneous nuclear ribonucleoprotein A0
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006805.3 1991 UTR 3 NP_006796.1

View Full Product Details