Product Details

SNP ID
rs1694089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:53951564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGATCACACAAATTTTCTAAAAGC[A/C]CTGATCTAGAGAACACAAGAATACC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARL15 PubMed Links

Gene Details

Gene
ARL15
Gene Name
ADP ribosylation factor like GTPase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019087.2 Intron NP_061960.1
XM_011543498.2 Intron XP_011541800.1
XM_011543499.2 Intron XP_011541801.1
XM_011543500.2 Intron XP_011541802.1
XM_017009598.1 Intron XP_016865087.1
Gene
MIR581
Gene Name
microRNA 581
There are no transcripts associated with this gene.

View Full Product Details