Product Details

SNP ID
rs1031744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:60756742 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAATCTTCACCACGCCATCTAAAC[A/C]AAATTCATACTTTCTCATTGGTGTT
Phenotype
MIM: 614451
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ELOVL7 PubMed Links

Gene Details

Gene
ELOVL7
Gene Name
ELOVL fatty acid elongase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104558.1 Intron NP_001098028.1
NM_001297617.1 Intron NP_001284546.1
NM_001297618.1 Intron NP_001284547.1
NM_024930.2 Intron NP_079206.2
XM_005248606.4 Intron XP_005248663.1
XM_005248607.3 Intron XP_005248664.1
XM_006714695.3 Intron XP_006714758.1
XM_011543651.2 Intron XP_011541953.1
XM_017009885.1 Intron XP_016865374.1
XM_017009886.1 Intron XP_016865375.1
XM_017009887.1 Intron XP_016865376.1
XM_017009888.1 Intron XP_016865377.1
XM_017009889.1 Intron XP_016865378.1

View Full Product Details