Product Details

SNP ID
rs1065265
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:66376216 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTTGATGAGGTTCACCATGGAAGA[C/T]GTAAAAGCTGTATCCCGAAGAAGGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00174 PubMed Links

Gene Details

Gene
LINC00174
Gene Name
long intergenic non-protein coding RNA 174
There are no transcripts associated with this gene.

View Full Product Details