Product Details

SNP ID
rs900614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72689812 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAACTGGGCTGCAGGCATGGAACA[A/T]TCAGGGCTGCAGACAGGGAGGAAAT
Phenotype
MIM: 606646
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARAP1 PubMed Links

Gene Details

Gene
ARAP1
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040118.2 Intron NP_001035207.1
NM_001135190.1 Intron NP_001128662.1
NM_015242.4 Intron NP_056057.2

View Full Product Details