Product Details

SNP ID
rs1451869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:64594769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAAGAGACATCTAAAGAAATTAC[A/G]TGGAGTGTAAGTTAGAGATACAAAG
Phenotype
MIM: 603711
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP7B1 PubMed Links

Gene Details

Gene
CYP7B1
Gene Name
cytochrome P450 family 7 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324112.1 3598 Intron NP_001311041.1
NM_004820.4 3598 UTR 3 NP_004811.1
XM_017014002.1 3598 UTR 3 XP_016869491.1

View Full Product Details