Product Details

SNP ID
rs1542644
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:106760163 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGACTCGTTATATGCTGACATGTAA[A/T]GAAACAATAAACTTCTCCAAGAAGT
Phenotype
MIM: 609747 MIM: 605609
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABRA PubMed Links

Gene Details

Gene
ABRA
Gene Name
actin binding Rho activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139166.4 2074 UTR 3 NP_631905.1
Gene
OXR1
Gene Name
oxidation resistance 1
There are no transcripts associated with this gene.

View Full Product Details