Product Details

SNP ID
rs3894197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124038988 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTGAGGATGAACGCTGTCAGTAGG[C/G]TGGCGTTGGACATTTCTTCTCATAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR10G7 PubMed Links

Gene Details

Gene
OR10G7
Gene Name
olfactory receptor family 10 subfamily G member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004463.1 14 Missense Mutation ACC,AGC T5S NP_001004463.1
Gene
OR10G8
Gene Name
olfactory receptor family 10 subfamily G member 8
There are no transcripts associated with this gene.

View Full Product Details