Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_173674.2 | 3851 | Intron | NP_775945.1 | ||
XM_005266941.1 | 3851 | Intron | XP_005266998.1 | ||
XM_006715463.1 | 3851 | Intron | XP_006715526.1 | ||
XM_011535771.1 | 3851 | Intron | XP_011534073.1 | ||
XM_011535773.2 | 3851 | Intron | XP_011534075.1 | ||
XM_011535774.2 | 3851 | Intron | XP_011534076.1 | ||
XM_017010775.1 | 3851 | Intron | XP_016866264.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001017408.2 | 3851 | UTR 3 | NP_001017408.1 | ||
NM_020399.3 | 3851 | UTR 3 | NP_065132.1 |