Product Details

SNP ID
rs1407076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116537154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACAGTTGTCTTACACTAGAGAAG[A/G]GTGAAGGTGATGAATATTCTCCTCA
Phenotype
MIM: 614137
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM26D PubMed Links

Gene Details

Gene
FAM26D
Gene Name
family with sequence similarity 26 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256887.2 Intron NP_001243816.1
NM_001256888.2 Intron NP_001243817.1
NM_001256889.2 Intron NP_001243818.1
NM_153036.4 Intron NP_694581.1
XM_005266860.3 Intron XP_005266917.1
XM_011535560.2 Intron XP_011533862.1
XM_011535561.2 Intron XP_011533863.1
XM_011535562.2 Intron XP_011533864.1
XM_011535563.2 Intron XP_011533865.1
XM_011535564.1 Intron XP_011533866.1
XM_017010390.1 Intron XP_016865879.1
XM_017010391.1 Intron XP_016865880.1
XM_017010392.1 Intron XP_016865881.1
Gene
TRAPPC3L
Gene Name
trafficking protein particle complex 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139444.2 Intron NP_001132916.1

View Full Product Details