Product Details
- SNP ID
-
rs1322772
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
25
- Location
-
Chr.1:173866620 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAGGTGTACTCTCTATGTTCCCCAA[A/C]TCACCATTTTGGGTGCCTCAGTTAA
- Phenotype
-
MIM: 610956
MIM: 608280
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DARS2
PubMed Links
Gene Details
- Gene
- DARS2
- Gene Name
- aspartyl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.
- Gene
- GAS5
- Gene Name
- growth arrest specific 5 (non-protein coding)
There are no transcripts associated with this gene.
- Gene
- GAS5-AS1
- Gene Name
- GAS5 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- SNORA103
- Gene Name
- small nucleolar RNA, H/ACA box 103
There are no transcripts associated with this gene.
- Gene
- SNORD44
- Gene Name
- small nucleolar RNA, C/D box 44
There are no transcripts associated with this gene.
- Gene
- SNORD47
- Gene Name
- small nucleolar RNA, C/D box 47
There are no transcripts associated with this gene.
- Gene
- SNORD74
- Gene Name
- small nucleolar RNA, C/D box 74
There are no transcripts associated with this gene.
- Gene
- SNORD75
- Gene Name
- small nucleolar RNA, C/D box 75
There are no transcripts associated with this gene.
- Gene
- SNORD76
- Gene Name
- small nucleolar RNA, C/D box 76
There are no transcripts associated with this gene.
- Gene
- SNORD77
- Gene Name
- small nucleolar RNA, C/D box 77
There are no transcripts associated with this gene.
- Gene
- SNORD78
- Gene Name
- small nucleolar RNA, C/D box 78
There are no transcripts associated with this gene.
- Gene
- SNORD79
- Gene Name
- small nucleolar RNA, C/D box 79
There are no transcripts associated with this gene.
- Gene
- SNORD80
- Gene Name
- small nucleolar RNA, C/D box 80
There are no transcripts associated with this gene.
- Gene
- SNORD81
- Gene Name
- small nucleolar RNA, C/D box 81
There are no transcripts associated with this gene.
- Gene
- ZBTB37
- Gene Name
- zinc finger and BTB domain containing 37
There are no transcripts associated with this gene.
View Full Product Details