Product Details

SNP ID
rs1043564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16518143 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCACCAGGCTGCCCATGGACACCA[C/G]GTGTGGCCGATTCCCAACGGTCACC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C19orf44 PubMed Links

Gene Details

Gene
C19orf44
Gene Name
chromosome 19 open reading frame 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288834.1 3841 UTR 3 NP_001275763.1
NM_032207.3 3841 Intron NP_115583.1
XM_006722920.3 3841 Intron XP_006722983.1
XM_011528354.2 3841 UTR 3 XP_011526656.1
XM_017027350.1 3841 Intron XP_016882839.1
Gene
CHERP
Gene Name
calcium homeostasis endoplasmic reticulum protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006387.5 3841 UTR 3 NP_006378.3

View Full Product Details