Product Details

SNP ID
rs1049601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17486723 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAATTCAAGGATGCGAAAACTAC[A/G]TCTATGACATAAACATGACATTCAA
Phenotype
MIM: 612676
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
QDPR PubMed Links

Gene Details

Gene
QDPR
Gene Name
quinoid dihydropteridine reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000320.2 1230 UTR 3 NP_000311.2
NM_001306140.1 1230 UTR 3 NP_001293069.1

View Full Product Details