Product Details

SNP ID
rs885701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31176123 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTCTGTCAACCACATCCCCACC[A/G]AAGTCACACTGCACCCGCTCTCCCT
Phenotype
MIM: 164177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POU5F1 PubMed Links

Gene Details

Gene
POU5F1
Gene Name
POU class 5 homeobox 1
There are no transcripts associated with this gene.

Gene
PSORS1C3
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details