Product Details

SNP ID
rs1265155
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:31175917 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGTCCCCAGCTGCTCCGGTTCCCCA[A/G]CAACTCACAGACAACCGTGGTCTGG
Phenotype
MIM: 164177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POU5F1 PubMed Links

Gene Details

Gene
POU5F1
Gene Name
POU class 5 homeobox 1
There are no transcripts associated with this gene.

Gene
PSORS1C3
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details