Product Details

SNP ID
rs1064799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151662495 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGTGAAACTCTTCACATCATGG[G/T]GAGAGTTTGTATGATTAATAAGAAG
Phenotype
MIM: 182120
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SPARC PubMed Links

Gene Details

Gene
SPARC
Gene Name
secreted protein acidic and cysteine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309443.1 2191 UTR 3 NP_001296372.1
NM_001309444.1 2191 UTR 3 NP_001296373.1
NM_003118.3 2191 UTR 3 NP_003109.1

View Full Product Details