Product Details

SNP ID
rs707156
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151661627 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCTCAAAATACGACACTAACAT[C/G]ATGAACATGCATGAGCTTTGAAAAG
Phenotype
MIM: 182120
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPARC PubMed Links

Gene Details

Gene
SPARC
Gene Name
secreted protein acidic and cysteine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309443.1 3059 UTR 3 NP_001296372.1
NM_001309444.1 3059 UTR 3 NP_001296373.1
NM_003118.3 3059 UTR 3 NP_003109.1

View Full Product Details