Product Details

SNP ID
rs2077694
Assay Type
Functionally tested
NCBI dbSNP Submissions
64
Location
Chr.1:89007451 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGATTTTTACCTAAGACTTTCCT[C/T]AGTATCCACTGGTCGTCTGGAAGAA
Phenotype
MIM: 600413
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GBP3 PubMed Links

Gene Details

Gene
GBP3
Gene Name
guanylate binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319179.1 2199 UTR 3 NP_001306108.1
NM_001319180.1 2199 UTR 3 NP_001306109.1
NM_001319181.1 2199 Intron NP_001306110.1
NM_018284.2 2199 UTR 3 NP_060754.2
XM_005270743.2 2199 UTR 3 XP_005270800.1
XM_006710574.2 2199 UTR 3 XP_006710637.1
XM_011541233.1 2199 UTR 3 XP_011539535.1
XM_011541234.1 2199 UTR 3 XP_011539536.1
XM_017001013.1 2199 UTR 3 XP_016856502.1
XM_017001014.1 2199 UTR 3 XP_016856503.1

View Full Product Details