Product Details

SNP ID
rs2305762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:34678635 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCAGGGAGAGATTGACGCTCCGG[A/C]GAGTGATTACCAGTGTGCTTTTCCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCGB1B2P PubMed Links

Gene Details

Gene
SCGB1B2P
Gene Name
secretoglobin family 1B member 2, pseudogene
There are no transcripts associated with this gene.

Gene
ZNF302
Gene Name
zinc finger protein 302
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012320.2 538 Intron NP_001012320.1
NM_001289181.1 538 Intron NP_001276110.1
NM_001289182.1 538 Intron NP_001276111.1
NM_001289183.1 538 Intron NP_001276112.1
NM_001289184.1 538 Intron NP_001276113.1
NM_001289185.1 538 Intron NP_001276114.1
NM_001289186.1 538 Intron NP_001276115.1
NM_001289187.1 538 Intron NP_001276116.1
NM_001289188.1 538 Intron NP_001276117.1
NM_001289189.1 538 Intron NP_001276118.1
NM_001289190.1 538 Intron NP_001276119.1
NM_001289191.1 538 Intron NP_001276120.1
NM_001289192.1 538 Intron NP_001276121.1
NM_018443.3 538 Intron NP_060913.2
NM_018675.2 538 Intron NP_061145.2
XM_011527111.2 538 Intron XP_011525413.1
XM_017026978.1 538 Intron XP_016882467.1
XM_017026979.1 538 Intron XP_016882468.1
XM_017026980.1 538 Intron XP_016882469.1
XM_017026981.1 538 Intron XP_016882470.1
XM_017026982.1 538 Intron XP_016882471.1
XM_017026983.1 538 Intron XP_016882472.1
XM_017026984.1 538 UTR 5 XP_016882473.1
XM_017026985.1 538 Intron XP_016882474.1
XM_017026986.1 538 Intron XP_016882475.1

View Full Product Details