Product Details

SNP ID
rs11578865
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:153153826 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATAATATGCCTTTAAGTTTTTGA[A/G]AATATAGAATTTAAAAATAATAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPRR2G PubMed Links

Gene Details

Gene
SPRR2G
Gene Name
small proline rich protein 2G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014291.3 Intron NP_001014313.1
XM_017002177.1 Intron XP_016857666.1
XM_017002178.1 Intron XP_016857667.1

View Full Product Details