Product Details

SNP ID
rs2053116
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:4803648 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCAAGATGACACAGATATGGGAGG[A/C]ACGTGTGCTAAAAGCTTTGAGGCGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR52R1 PubMed Links

Gene Details

Gene
OR52R1
Gene Name
olfactory receptor family 52 subfamily R member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005177.3 733 Missense Mutation GCC,TCC A245S NP_001005177.3

View Full Product Details