Product Details

SNP ID
rs4759322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54071880 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCATTAAAGCACATGGTGTTAAGT[A/G]CAGTCCTCTGCAGATCACATCCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLJ12825 PubMed Links

Gene Details

Gene
FLJ12825
Gene Name
uncharacterized LOC440101
There are no transcripts associated with this gene.

Gene
LOC100240735
Gene Name
uncharacterized LOC100240735
There are no transcripts associated with this gene.

View Full Product Details